Skip to Content
Merck
All Photos(1)

Key Documents

903477

Sigma-Aldrich

Androsterone-2,2,4,4-d4 3-glucronide sodium salt

≥98 atom % D, ≥98% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€472.00

€472.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 MG
€472.00

About This Item

Empirical Formula (Hill Notation):
C25D4H33O8Na
Molecular Weight:
492.57
UNSPSC Code:
12352106
NACRES:
NA.24

€472.00


Check Cart for Availability

Request more information

isotopic purity

≥98 atom % D

Quality Level

Assay

≥98% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mass shift

M+4

storage temp.

−20°C

SMILES string

[2H]C1(C[C@@]2(C3CC[C@]4(C)C(CCC4C3CC[C@]2(C([2H])([2H])[C@@H]1O[C@@H]5O[C@@H]([C@H](C([C@H]5O)O)O)C([O-])=O)[H])=O)C)[2H].[Na+]

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service