Skip to Content
Merck
All Photos(3)

Key Documents

903124

Sigma-Aldrich

(Ad−BippyPhos)2PdCl2

Synonym(s):

Bis[5-(di(1-adamantyl)phosphino)-1′,3′,5′-triphenyl-1′H-[1,4′]bipyrazole] palladium(II) dichloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€112.00
250 MG
€205.00

€112.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€112.00
250 MG
€205.00

About This Item

Empirical Formula (Hill Notation):
C88H94N8P2PdCl2
Molecular Weight:
1503.02
UNSPSC Code:
12161600
NACRES:
NA.22

€112.00


Check Cart for Availability

Request a Bulk Order

form

powder or crystals

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

mp

263 °C (Decomp)

functional group

phosphine

Application

A Pd based catalyst with bulky ligand applicable to various cross-coupling reactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service