Skip to Content
Merck
All Photos(1)

Key Documents

902950

Sigma-Aldrich

Conductive ink for solvent-cast 3D printers

for layer-by-layer 3D structure fabrication

Synonym(s):

30 wt% Carbon nanotubes nanocomposite ink, BlackElectron30-A0 ink

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002

description

Solid content: 10 wt% solid (CNT/PLA nanocomposite) in dichloromethane (30 wt% of CNT in solid part)

form

liquid

SMILES string

OC(C1=C2C(C3=C(C4=C(C5=C(C6=C(C7=C(C8=C(C9=C(C%10=C(C%11=C(C%12=C(C%13=C(C%14=C(C%15=CC=C%16O)C%16=C%17)C%17=C%18)C%18=C%19C(O)=O)C%19=C%20)C%20=C%21O)C%21=C%22)C%22=C%23)C%23=C%24O)C%24=C%25)C%25=C%26C(O)=O)C%26=C%27)C%27=C2)=C%28C%29=C(C%30=C%31C%28=C%3

Looking for similar products? Visit Product Comparison Guide

Legal Information

Sold under Material Transfer Agreement with Daniel Therriault group at Polytechnique Montreal.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rouhollah D Farahani et al.
Advanced materials (Deerfield Beach, Fla.), 28(28), 5794-5821 (2016-05-03)
The integration of nanotechnology into three-dimensional printing (3DP) offers huge potential and opportunities for the manufacturing of 3D engineered materials exhibiting optimized properties and multifunctionality. The literature relating to different 3DP techniques used to fabricate 3D structures at the macro-
Shuang-Zhuang Guo et al.
Small (Weinheim an der Bergstrasse, Germany), 9(24), 4118-4122 (2013-07-05)
The solvent-cast direct-write fabrication of microstructures is shown using a thermoplastic polymer solution ink. The method employs the robotically controlled microextrusion of a filament combined with a rapid solvent evaporation. Upon drying, the increased rigidity of the extruded filament enables

Articles

Graphene nanoribbons (GNRs) are quasi-one-dimensional narrow strips of graphene comprised of sp2-hybridized carbon atoms arranged into hexagonal honeycomb lattice configurations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service