Skip to Content
Merck
All Photos(2)

Key Documents

902829

Sigma-Aldrich

2-(2,4-Difluorophenyl)-5-methylpyridine

≥95%

Synonym(s):

Photocatalysis ligand

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9F2N
CAS Number:
Molecular Weight:
205.20
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

Assay

≥95%

form

solid

reaction suitability

reagent type: catalyst
reagent type: ligand
reaction type: Photocatalysis

Application

This ligand is used in Ir(III) complexes suitable for use in photocatalysis and phosphorescent OLEDs.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of substituents on properties of diphenylphosphoryl-substituted bis-cyclometalated Ir(III) complexes with a picolinic acid as ancillary ligand.
Yu H, et al.
Dyes and Pigments (2017)
Chip Le et al.
Science (New York, N.Y.), 360(6392), 1010-1014 (2018-06-02)
Transition metal-catalyzed arene functionalization has been widely used for molecular synthesis over the past century. In this arena, copper catalysis has long been considered a privileged platform due to the propensity of high-valent copper to undergo reductive elimination with a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service