Skip to Content
Merck
All Photos(1)

Key Documents

902772

Sigma-Aldrich

(Diacetoxyiodo)benzene solution

0.50 M solution in DCM

Synonym(s):

Iodobenzene diacetate, Iodosobenzene diacetate, Phenyl-iodanediyl diacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H11IO4
CAS Number:
MDL number:
UNSPSC Code:
12352002

form

liquid

reaction suitability

reaction type: C-H Activation
reagent type: catalyst
reagent type: oxidant

refractive index

n/D 1.444

density

1.361 g/mL

InChI

1S/C10H11IO4/c1-8(12)14-11(15-9(2)13)10-6-4-3-5-7-10/h3-7H,1-2H3

InChI key

ZBIKORITPGTTGI-UHFFFAOYSA-N

Application

Stoichiometric oxidant in the TEMPO oxidation of nerol to neral.[1] Oxidant employed in the rhodium-catalyzed aziridination of olefins with sulfamate esters.[2]
Unactivated sp3 C-H bonds of both oxime and pyridine substrates undergo highly regio- and chemoselective Pd(II)-catalyzed oxygenation with PhI(OAc)2 as a stoichiometric oxidant.[3]
Used in the room temperature Pd-catalyzed 2-arylation of indoles
Useful reagent for the synthesis of a wide variety of heterocyclic compounds.[4][5]

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jayasree Seayad et al.
Organic letters, 12(7), 1412-1415 (2010-03-13)
Copper(I) or -(II) salts with weakly coordinating anions catalyze the diacetoxylation of olefins efficiently in the presence of PhI(OAc)(2) as the oxidant under mild conditions. The reaction is effective for aryl, aryl alkyl, as well as aliphatic terminal and internal
Synlett, 221-221 (1994)
Substrate dependence of nonlinear effects: mechanistic probe and practical applications.
Y K Chen et al.
Journal of the American Chemical Society, 123(22), 5378-5379 (2001-07-18)
Hai Yi et al.
Chemical communications (Cambridge, England), 46(37), 6941-6943 (2010-08-24)
3-Aryl-4-unsubstituted-6-CF(3)-pyridin-2-ones have been efficiently synthesized from readily available 4-aryl-3-carbamoyl-6-CF(3)-pyridin-2(1H)-ones by treatment with PhI(OAc)(2) in the presence of NaOH.
Aldrichimica Acta, 27, 15-15 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service