Skip to Content
Merck
All Photos(1)

Key Documents

902675

Sigma-Aldrich

BocNH-PEG6-acid

Synonym(s):

2,2-Dimethyl-4-oxo-3,8,11,14,17,20,23-heptaoxa-5-azapentacosan-25-oic acid, Boc-NH-PEG6-CH2COOH

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€248.00
1 G
€763.00

€248.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€248.00
1 G
€763.00

About This Item

Empirical Formula (Hill Notation):
C19H37NO10
CAS Number:
Molecular Weight:
439.50
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22

€248.00


Check Cart for Availability

Request a Bulk Order

form

liquid

reaction suitability

reagent type: linker

refractive index

n/D 1.4657

functional group

Boc
amine
carboxylic acid

storage temp.

−20°C

SMILES string

OC(COCCOCCOCCOCCOCCOCCNC(OC(C)(C)C)=O)=O

InChI

1S/C19H37NO10/c1-19(2,3)30-18(23)20-4-5-24-6-7-25-8-9-26-10-11-27-12-13-28-14-15-29-16-17(21)22/h4-16H2,1-3H3,(H,20,23)(H,21,22)

InChI key

ZKCDWMFMVAYYED-UHFFFAOYSA-N

Application

This heterobifunctional, PEGylated crosslinker features a carboxylic acid at one end and Boc-protected amino group at the other, which can be deprotected with mildly acidic conditions. The hydrophillic PEG linker facilitates solubility in biological applications. BocNH-PEG6-acid can be used for bioconjugation or as a building block for synthesis of small molecules, conjugates of small molecules and/or biomolecules, or other tool compounds for chemical biology and medicinal chemistry that require ligation. Examples of applications include its synthetic incorporation into antibody-drug conjugates or proteolysis-targeting chimeras (PROTAC® molecules) for targeted protein degradation.

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vincent Rerat et al.
Journal of medicinal chemistry, 52(22), 7029-7043 (2009-10-29)
RGD peptides are used in biomaterials science for surface modifications with a view to elicit selective cellular responses. Our objective is to replace peptides by small peptidomimetics acting similarly. We designed novel molecules targeting alpha(v)beta(3) integrin and featuring spacer-arms (for
Vincent Rerat et al.
Bioorganic & medicinal chemistry letters, 20(6), 1861-1865 (2010-02-23)
Ultrasmall particles of iron oxide (USPIOs) coated with 3,3'-bis(phosphonate)propionic acid were covalently coupled to a home-made Arg-Gly-Asp (RGD) peptidomimetic molecule via a short oligoethylene-glycol (OEG) spacer. The conjugation rate was measured by X-ray photoelectron spectroscopy (XPS). The particle size and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service