Skip to Content
Merck
All Photos(1)

Key Documents

902543

Sigma-Aldrich

Propargyl-PEG4-acid

Synonym(s):

4,7,10,13-Tetraoxahexadec-15-ynoic acid, Propargyl-PEG4-CH2CO2H

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H20O6
CAS Number:
Molecular Weight:
260.28
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22
Pricing and availability is not currently available.

form

liquid

reaction suitability

reagent type: linker

availability

available only in USA

refractive index

n/D 1.460

density

1.164 g/mL

functional group

alkyne
carboxylic acid

storage temp.

−20°C

SMILES string

OC(CCOCCOCCOCCOCC#C)=O

InChI key

WMBXAUWIPBFEOK-UHFFFAOYSA-N

Application

This heterobifunctional, PEGylated crosslinker features a carboxylic acid at one end and propargyl group at the other for reaction with azide-containing compounds using click chemistry. The hydrophillic PEG linker facilitates solubility in biological applications. Propargyl-PEG4-acid can be used for bioconjugation or as a building block for synthesis of small molecules, conjugates of small molecules and/or biomolecules, or other tool compounds for chemical biology and medicinal chemistry that require ligation. Examples of applications include its synthetic incorporation into antibody-drug conjugates or proteolysis-targeting chimeras (PROTAC® molecules) for targeted protein degradation.

Technology Spotlight: Degrader Building Blocks for Targeted Protein Degradation

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

related product

Product No.
Description
Pricing

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Debabrata Bhunia et al.
Archiv der Pharmazie, 348(10), 689-703 (2015-09-04)
A Cu-mediated azide-alkyne click chemistry protocol was employed for the synthesis of a focused library of novel 1,2,3-triazolyl conjugates bearing various carbohydrate-steroid/triterpenoid entities. The immunogenicity of these compounds was examined initially by ex vivo assays. The lead compound 15g was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service