Skip to Content
Merck
All Photos(1)

Key Documents

902535

Sigma-Aldrich

Amino-PEG4-t-butyl ester

Synonym(s):

tert-Butyl 1-amino-3,6,9,12-tetraoxapentadecan-15-oate, H2N-PEG4-CH2CH2COOtBu, H2N-dPEG(4)-CO-OtBu

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€160.00
1 G
€509.00

€160.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€160.00
1 G
€509.00

About This Item

Empirical Formula (Hill Notation):
C15H31NO6
CAS Number:
Molecular Weight:
321.41
MDL number:
UNSPSC Code:
51171641
NACRES:
NA.22

€160.00


Check Cart for Availability

Request a Bulk Order

form

liquid

reaction suitability

reagent type: cross-linking reagent

refractive index

n/D 1.4508

density

1.04479 g/mL

functional group

amine
ester

storage temp.

−20°C

SMILES string

NCCOCCOCCOCCOCCC(OC(C)(C)C)=O

InChI

1S/C15H31NO6/c1-15(2,3)22-14(17)4-6-18-8-10-20-12-13-21-11-9-19-7-5-16/h4-13,16H2,1-3H3

InChI key

PKESARRNSGIDRD-UHFFFAOYSA-N

Application

This heterobifunctional, PEGylated crosslinker features an amino group at one end and t-butyl-protected carboxyl group at the other, which can be deprotected with acidic conditions. The hydrophilic PEG linker facilitates solubility in biological applications. Amino-PEG4-t-butyl ester can be used for bioconjugation or as a building block for synthesis of small molecules, conjugates of small molecules and/or biomolecules, or other tool compounds for chemical biology and medicinal chemistry that require ligation. Examples of applications include its synthetic incorporation into antibody-drug conjugates or proteolysis-targeting chimeras (PROTAC® molecules) for targeted protein degradation.

Technology Spotlight: Degrader Building Blocks for Targeted Protein Degradation

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service