Skip to Content
Merck
All Photos(3)

Key Documents

902136

Sigma-Aldrich

2,4,6-Tri-(4-fluorophenyl)pyrylium tetrafluoroborate

≥95%

Synonym(s):

Triphenylpyrylium photosensitizer, [T(p-F)PPT

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€116.00

€116.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€116.00

About This Item

Empirical Formula (Hill Notation):
C23H14BF7O
CAS Number:
Molecular Weight:
450.16
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

€116.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95%

form

powder

reaction suitability

reagent type: catalyst
reaction type: Photocatalysis

mp

242-245 °C

photocatalyst activation

465 nm

SMILES string

[B-](F)(F)(F)F.Fc1ccc(cc1)c2[o+]c(cc(c2)c4ccc(cc4)F)c3ccc(cc3)F

InChI key

FRADIBPDZCOIFV-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kuai Wang et al.
Organic letters, 19(8), 1958-1961 (2017-04-04)
A highly regioselective [2 + 2 + 2] cyclization of aromatic alkynes with nitriles is developed for the preparation of 2,3,6-trisubstituted pyridines under visible-light irradiation using a pyrylium salt as the photoredox catalyst. This cycloaddition is achieved through a photooxidative
Antonio Franconetti et al.
Physical chemistry chemical physics : PCCP, 16(34), 18442-18453 (2014-07-30)
Noncovalent interactions of anions with electron-deficient aromatic rings that have been studied so far involve non-heteroaromatic or nitrogen-based heteroaromatic systems. Here we report the first case of an organic oxygenated aromatic system, in particular the tri-aryl-pyrylium tetrafluoroborate system, for which
Photocatalytic Cross-Dehydrogenative Amination Reactions between Phenols and Diarylamines.
Zhao Y, et al.
ACS Catalysis, 7 (4), 2446-2451 (2017)
Ji Young Cho et al.
The Journal of organic chemistry, 83(2), 805-811 (2017-12-14)
Dibenzofurans are naturally occurring molecules that have received considerable attention for a variety of practical applications, such as in pharmaceuticals and electronic materials. Herein, an efficient and eco-friendly method for the synthesis of dibenzofuran derivatives via intramolecular C-O bond formation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service