Skip to Content
Merck
All Photos(1)

Key Documents

901797

Sigma-Aldrich

Molybdenum disulfide suspension

greener alternative

Synonym(s):

2D-Moly sulfide dispersion, 2D-Molybdenum disulfide dipsersion with nonionic surfactant, 2D-Molybdenum disulfide dispersion, Moly sulfide

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
€247.00

€247.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 ML
€247.00

About This Item

Linear Formula:
MoS2
UNSPSC Code:
12352300
NACRES:
NA.23

€247.00


Check Cart for Availability

Request a Bulk Order

description

50-1000 nm
thickness < 3 layers

form

liquid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

1 mg/mL in H2O

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

Molybdenum disulfide suspension (MoS2) is a two dimensional (2D) transition metal dichalcogenide (TMD) material that forms a monolayer with a direct band gap of 1.8 eV and an indirect band gap of 1.2 eV. It can be used as a non-linear optical material and as a reinforcing filler in polymeric composites.[1][2][3]
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". Click here for more information.

Application

MoS2 suspension can be used in the fabrication of semiconducting devices such as field effect transistors (FETs), and supercapacitors.[1][4]
Owing to the thickness dependent physical and chemical properties, the transition metal dichalcogenide (TMD) based two dimensional (2D) nanomaterials have drawn significant attention recently.[5][6][7][8] The atomically thin layers of MoS2 and WS2 represent important class of TMD based 2D-material and they find applications in photocatalysis, phtotovoltaics, gas sensing, lithium ion batteries, field effect transistors, spintronics etc.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The super materials that could trump graphene.
Gibney E
Nature, 522(7556), 274-276 (2015)
Annealing and transport studies of suspended molybdenum disulfide devices.
Wang F, et al.
Nanotechnology, 26(10), 105709-105709 (2015)
Supercapacitive energy storage performance of molybdenum disulfide nanosheets wrapped with microporous carbons.
Weng Q, et al.
Journal of Material Chemistry A, 3(6), 3097-3102 (2015)
Exploration of Nanostructured Functional Materials Based on Hybridization of Inorganic 2D Nanosheets.
Gunjakar J L, et al
The Journal of Physical Chemistry C, 118, 3847- 3847 (2014)
Structure and catalytic properties of molybdenum sulfide nanoplatelets.
Elizondo-Villarreal N, et al.
Applied Catalysis A: General, 328(1), 88-97 (2007)

Articles

Recent advancements in paper-based sensing platforms offer cost-effective clinical diagnostics with microfluidic channels and colorimetric or electrochemical detection zones.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service