Skip to Content
Merck
All Photos(1)

Key Documents

901770

Sigma-Aldrich

Nanodiamonds

65 nm octadecane functionalized, powder

Synonym(s):

Detonation nanodiamond, Diamond nanoparticles, Nanodiamonds

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€414.00

€414.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€414.00

About This Item

Linear Formula:
C
UNSPSC Code:
12352100
NACRES:
NA.23

€414.00


Check Cart for Availability

Request a Bulk Order

form

nanoparticles
powder

Looking for similar products? Visit Product Comparison Guide

Application

Nanodiamonds modified with hydrocarbons exhibit colloidal stability in hydrophobic and non-polar solvents including lithium ion battery electrolytes. Hydrocarbon modified nanodiamonds have also been shown to suppress the dendrite formation in lithium ion batteries[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin-Bing Cheng et al.
Nature communications, 8(1), 336-336 (2017-08-26)
Lithium metal has been regarded as the future anode material for high-energy-density rechargeable batteries due to its favorable combination of negative electrochemical potential and high theoretical capacity. However, uncontrolled lithium deposition during lithium plating/stripping results in low Coulombic efficiency and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service