Skip to Content
Merck
All Photos(2)

Key Documents

901749

Sigma-Aldrich

(R)-5-((1H-1,2,3-Triazol-1-yl)methyl)-3-(3-fluoro-4-iodophenyl)oxazolidin-2-one

Synonym(s):

Aryl halide chemistry informer library compound X14

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€143.00

€143.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€143.00

About This Item

Empirical Formula (Hill Notation):
C12H10FIN4O2
CAS Number:
Molecular Weight:
388.14
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

€143.00


Check Cart for Availability

Request a Bulk Order

form

powder or crystals

mp

195-200 °C (w/ decomp.)

Application

This product is Informer compound X14 of the Aryl halide chemistry informer library developed by chemists at Merck & Co., Inc., Kenilworth, NJ, U.S., which contains 18 drug-like molecules representative of those encountered in complex synthesis. By screening a new reaction against the informer library, chemists can directly compare and analyze a reactions′s successes and shortcomings among different methods and various research teams. It may also be used to facilitate deeper method development for performance or utility.

Caution

Not fully tested.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nanomole-scale high-throughput chemistry for the synthesis of complex molecules.
Buitrago Santanilla, et al.
Science, 347, 49-53 (2015)
Alexander Buitrago Santanilla et al.
Science (New York, N.Y.), 347(6217), 49-53 (2015-01-03)
At the forefront of new synthetic endeavors, such as drug discovery or natural product synthesis, large quantities of material are rarely available and timelines are tight. A miniaturized automation platform enabling high-throughput experimentation for synthetic route scouting to identify conditions
Emily B Corcoran et al.
Science (New York, N.Y.), 353(6296), 279-283 (2016-06-25)
Over the past two decades, there have been major developments in transition metal-catalyzed aminations of aryl halides to form anilines, a common structure found in drug agents, natural product isolates, and fine chemicals. Many of these approaches have enabled highly
Peter S Kutchukian et al.
Chemical science, 7(4), 2604-2613 (2016-04-21)
Major new advances in synthetic chemistry methods are typically reported using simple, non-standardized reaction substrates, and reaction failures are rarely documented. This makes the evaluation and choice of a synthetic method difficult. We report a standardized complex molecule diagnostic approach

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service