Skip to Content
Merck
All Photos(1)

Key Documents

901681

Sigma-Aldrich

Phosphonic acid-18O3

≥95 atom % 18O, ≥98% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€1,380.00

€1,380.00


Check Cart for Availability

Request more information

Select a Size

Change View
10 MG
€1,380.00

About This Item

Empirical Formula (Hill Notation):
H318O3P
CAS Number:
Molecular Weight:
88.00
MDL number:
UNSPSC Code:
12352301
NACRES:
NA.12

€1,380.00


Check Cart for Availability

Request more information

isotopic purity

≥95 atom % 18O

Quality Level

Assay

≥98% (CP)

form

solid

mass shift

M+2

storage temp.

room temp

SMILES string

[18OH]P([18OH])([H])=[18O]

InChI

1S/H3O3P/c1-4(2)3/h4H,(H2,1,2,3)/i1+2,2+2,3+2

InChI key

ABLZXFCXXLZCGV-FIXSIXJRSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service