Skip to Content
Merck
All Photos(1)

Key Documents

901623

Sigma-Aldrich

Buffered oxide etchant (BOE) 6:1 with surfactant

Synonym(s):

BHF, Buffered HF

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€106.00

€106.00


Estimated to ship on03 June 2025


Request a Bulk Order

Select a Size

Change View
1 L
€106.00

About This Item

UNSPSC Code:
12161700
NACRES:
NA.23

€106.00


Estimated to ship on03 June 2025


Request a Bulk Order

form

liquid

Related Categories

General description

Buffered oxide etchant (BOE) is a wet etchant used in microfabrication. Its primary use is in etching thin films of silicon dioxide (SiO2) or silicon nitride (Si3N4). It is a mixture of a buffering agent, such as ammonium fluoride (NH4F), and hydrofluoric acid (HF). Concentrated HF etches silicon dioxide too quickly for good process control and also peels photoresist used in lithographic patterning.

Application

Buffered oxide etchant (BOE) 6:1 with surfactant may be used in the oxide removal of AlGaN/GaN-based high electron mobility transistors for gate photolithography.[1][2] It may also be used in a buffer oxide etchant method for the fabrication of micro biochip.[3]

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of passivation on AlGaN/GaN HEMT device performance
2000 IEEE international symposium on compound semiconductors, 357-363 (2000)
Fabrication of a new micro bio chip and flow cell cytometry system using Bio-MEMS technology
Byun I, et al.
Microelectronics Journal, 39(5), 717-722 (2008)
Surface cleaning and preparation in AlGaN/GaN-based HEMT processing as assessed by X-ray photoelectron spectroscopy
Gonzalez-Posada F, et al.
Applied Surface Science, 253(14), 6185-6190 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service