Skip to Content
Merck
All Photos(1)

Key Documents

901296

Sigma-Aldrich

rac-Timolol-1,1,2,3,3-d5 maleate

≥98 atom % D, ≥98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13D5H19N4O3S · C4H4O4
CAS Number:
Molecular Weight:
437.52
MDL number:
UNSPSC Code:
12352005

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

≥98 atom % D

Assay

≥98% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mass shift

M+5

storage temp.

−20°C

SMILES string

CC(C)(C)NC([2H])([2H])C(O)([2H])C([2H])([2H])OC1=NSN=C1N2CCOCC2.OC(/C=C\C(O)=O)=O

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service