Skip to Content
Merck
All Photos(1)

Key Documents

901132

Sigma-Aldrich

1-Azido-2-(2-(2-methoxyethoxy)ethoxy)ethane

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€270.00

€270.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€270.00

About This Item

Empirical Formula (Hill Notation):
C7H15N3O3
CAS Number:
Molecular Weight:
189.21
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

€270.00


Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

reaction suitability

reaction type: click chemistry

refractive index

n/D 1.4472

density

1.08 g/mL

storage temp.

2-8°C

SMILES string

COCCOCCOCCN=[N+]=[N-]

InChI

1S/C7H15N3O3/c1-11-4-5-13-7-6-12-3-2-9-10-8/h2-7H2,1H3

InChI key

ZSSKNHYYIBAWRX-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service