Skip to Content
Merck
All Photos(2)

Key Documents

901104

Sigma-Aldrich

Hex-Ir(piq)3

Synonym(s):

Tris[2-(4-n-hexylphenyl)quinoline)]iridium(III)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€290.00

€290.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 MG
€290.00

About This Item

Empirical Formula (Hill Notation):
C63H66IrN3
CAS Number:
Molecular Weight:
1057.44
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

€290.00


Estimated to ship on28 May 2025


Request a Bulk Order

Assay

≥95%

Quality Level

form

solid

color

red

SMILES string

CCCCCCC1=CC([Ir+3](C2=C(C3=C(C=CC=C4)C4=CC=N3)C=CC(CCCCCC)=C2)C5=C(C6=C(C=CC=C7)C7=CC=N6)C=CC(CCCCCC)=C5)=C(C8=C(C=CC=C9)C9=CC=N8)C=C1

InChI

1S/3C21H22N.Ir/c3*1-2-3-4-5-8-17-11-13-19(14-12-17)21-20-10-7-6-9-18(20)15-16-22-21;/h3*6-7,9-13,15-16H,2-5,8H2,1H3;/q;;;+3

InChI key

FHDHOHBBTMTVSN-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service