Skip to Content
Merck
All Photos(1)

Key Documents

900979

Sigma-Aldrich

F8BT

average Mw >20,000

Synonym(s):

Poly(9,9-dioctylfluorene-alt-benzothiadiazole), Poly[(9,9-di-n-octylfluorenyl-2,7-diyl)-alt-(benzo[2,1,3]thiadiazol-4,8-diyl)]

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€399.00

€399.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€399.00

About This Item

Linear Formula:
(C35H42N2S)n
CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

€399.00


Check Cart for Availability

Request a Bulk Order

form

solid

mol wt

average Mn >20,000
average Mw >20,000

color

, Light Brown to Very Dark Brown to Very Dark Orange

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tuning the Emission of Semiconducting Polymer Dots from Green to Near-Infrared by Alternating Donor Monomers and Their Applications for in Vivo Biological Imaging.
Liou SY, et al.
ACS Macro Letters, 5(1), 154-157 (2016)
Xu Zhen et al.
ACS nano, 10(6), 6400-6409 (2016-06-15)
Detection of reactive oxygen species (ROS), a hallmark of many pathological processes, is imperative to understanding, detection and treatment of many life-threatening diseases. However, methods capable of real-time in situ imaging of ROS in living animals are still very limited.
Shouying Li et al.
ACS applied materials & interfaces, 8(6), 3624-3634 (2015-10-23)
This paper described the energy-transfer amplified singlet oxygen generation in semiconductor polymer dots (Pdots) for in vitro and in vivo photodynamic therapy. Hydrophobic photosensitizer tetraphenylporphyrin was facilely doped in the nanoparticles consisting of densely packed semiconductor polymers. Optical characterizations indicated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service