Skip to Content
Merck
All Photos(2)

Key Documents

900937

Sigma-Aldrich

TBPe

greener alternative

sublimed

Synonym(s):

2,5,8,11-Tetra-tert-butylperylene, 2,5,8,11-Tetrakis(1,1-dimethylethyl)perylene, TBP

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€316.00

€316.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€316.00

About This Item

Empirical Formula (Hill Notation):
C36H44
CAS Number:
Molecular Weight:
476.73
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

€316.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99% (sublimed, HPLC)

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

SMILES string

C(C)(C)(C)c1cc2c3c(c4c5c2cc(cc5cc(c4)C(C)(C)C)C(C)(C)C)cc(cc3c1)C(C)(C)C

InChI

1S/C36H44/c1-33(2,3)23-13-21-14-24(34(4,5)6)19-29-30-20-26(36(10,11)12)16-22-15-25(35(7,8)9)18-28(32(22)30)27(17-23)31(21)29/h13-20H,1-12H3

InChI key

BFTIPCRZWILUIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Electron transport and hole blocking material in organic light emitting diodes (OLED).
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". Electron transport organic materials have the chemical functionality to transport electrons and are used as electron transport layer in OLED devices. Click here for more information.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

High efficiency blue fluorescent organic light-emitting diodes using a conventional blue fluorescent emitter.
Lee IH, et al.
Journal of Material Chemistry C, 3(34), 8834-8838 (2015)
Wook Song et al.
Advanced materials (Deerfield Beach, Fla.), 27(29), 4358-4363 (2015-06-17)
High quantum efficiency in blue and white fluorescence organic light-emitting diodes is achieved by developing a novel device architecture with fluorescent emitters doped in a thermally activated delayed fluorescent emitter as a host material.
Tetradentate Pt(II) complexes with 6-membered chelate rings: A new route for stable and efficient blue organic light Emitting Diodes.
Fleetham TB, et al.
Chemistry of Materials, 28(10), 3276-3282 (2016)

Articles

Explore the eco-friendly potential of organic thin film transistors (OTFTs) for detecting chemical analytes, identifying viruses, and assisting in health diagnostics. This mini-review highlights challenges of achieving sustainability, safety, and biodegradability of each component of an OTFT sensor.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service