Skip to Content
Merck
All Photos(2)

Key Documents

900871

Sigma-Aldrich

2,2-Dimethyl-3,6,9,12-tetraoxa-2-silatridecane

97%, anhydrous

Synonym(s):

ANL-1NM3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H24O4Si
CAS Number:
Molecular Weight:
236.38
MDL number:
UNSPSC Code:
26111700
NACRES:
NA.23

grade

battery grade

Assay

97%

form

liquid

quality

anhydrous

impurities

≤500 ppm H2O

bp

240 °C

density

0.927 g/cm3

application(s)

battery manufacturing

SMILES string

C[Si](C)(C)OCCOCCOCCOC

InChI

1S/C10H24O4Si/c1-11-5-6-12-7-8-13-9-10-14-15(2,3)4/h5-10H2,1-4H3

InChI key

LGPOIKHOZYPSME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

In lithium ion batteries (LIBs) the redox shuttles are employed to prevent overcharging which could lead to thermal runaway and catastrophic failure. The use of redox shuttles increases the safety and stability of LIBs. 1, 4-di-tert-butyl-2, 5-bis(2-methoxyethoxy) benzene is an important redox shuttle used for overcharge protection in batteries.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

215.6 °F

Flash Point(C)

102 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barbarich TJ and Patterson ML
ECS Transactions, 45, 49-49 (2013)
Zhang L, et al.
ECS Transactions, 45, 57-57 (2013)
An All-Organic Non-aqueous Lithium-Ion Redox Flow Battery.
Brushett FR, et al.
Advanced Energy Materials, 2, 1390-1396 (2012)

Articles

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Ionic liquid electrolytes explored for rechargeable batteries' advancement; future IL development discussed.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service