Skip to Content
Merck
All Photos(2)

Documents

900584

Sigma-Aldrich

5-Ethynyl-2′-deoxyuridine

95%

Synonym(s):

EdU

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N2O5
CAS Number:
Molecular Weight:
252.22
UNSPSC Code:
12352208
NACRES:
NA.22

Quality Level

Assay

95%

form

powder

mp

206 °C

storage temp.

−20°C

SMILES string

OC[C@@H]1[C@@H](O)C[C@H](N2C(NC(C(C#C)=C2)=O)=O)O1

InChI

1S/C11H12N2O5/c1-2-6-4-13(11(17)12-10(6)16)9-3-7(15)8(5-14)18-9/h1,4,7-9,14-15H,3,5H2,(H,12,16,17)/t7-,8+,9+/m0/s1

InChI key

CDEURGJCGCHYFH-DJLDLDEBSA-N

General description

5-Ethynyl-2′-deoxyuridine (EdU) is a thymidine analogue that can be incorporated into cellular DNA for cell proliferation studies. The incorporated nucleoside analogue can be detected by a copper-catalyzed click reaction with a fluorescent azide.

Application

5-Ethynyl-2′-deoxyuridine (EdU) is used for:
  • Labeling newly synthesized DNA during replication
  • Cell proliferating assay studies
  • Cell cycle analysis
For information on Baseclick kits containing all the reagents for the assays see: Baseclick kits

For a listing of the Baseclick kits see: Baseclick kits

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Muta. 1B - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, susceptibility to enzymatic phosphorylation, cytotoxicity and in vitro antiviral activity of lipophilic pyrimidine nucleoside/carborane conjugates.
Bialek-Pietras M, et al.
Journal of Organometallic Chemistry, 865, 166-172 (2018)
Evaluation of 5-ethynyl-2?-deoxyuridine staining as a sensitive and reliable method for studying cell proliferation in the adult nervous system
C Zeng, et.al.
Brain Research, 1319, 21-32 (2010)
5-Ethynyl-2?-deoxycytidine as a new agent for DNA labeling: detection of proliferating cells
D Qu, et.al.
Analytical Biochemistry, 417, 112-121 (2011)
Cell type specific applicability of 5-ethynyl-2?-deoxyuridine (EdU) for dynamic proliferation assessment in flow cytometry
S Diermeier
Cytometry. Part A : the Journal of the International Society For Analytical Cytology, 75, 535-546 (2009)
Site-directed spin-labeling of nucleic acids by click chemistry: detection of abasic sites in duplex DNA by EPR spectroscopy.
Jakobsen A, et al.
Journal of the American Chemical Society, 132(30), 10424-10428 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service