Skip to Content
Merck
All Photos(2)

Key Documents

900472

Sigma-Aldrich

(R)-TolBINAP Pd G4

95%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€622.00

€622.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€622.00

About This Item

Empirical Formula (Hill Notation):
C62H55NO3P2PdS
Molecular Weight:
1062.54
UNSPSC Code:
12161600
NACRES:
NA.22

€622.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

powder

feature

generation 4

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

functional group

phosphine

SMILES string

CC(C=C1)=CC=C1P(C2=CC=C(C)C=C2)C3=C(C4=C(P(C5=CC=C(C)C=C5)C6=CC=C(C)C=C6)C=CC7=C4C=CC=C7)C8=C(C=C3)C=CC=C8.C[NH+](C9=C(C=CC=C9)C%10=C%11C=CC=C%10)[Pd-]%11OS(C)(=O)=O

InChI key

WKGRPDDVBKRQJX-UHFFFAOYSA-M

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathaniel H Park et al.
Angewandte Chemie (International ed. in English), 54(28), 8259-8262 (2015-06-03)
In Pd-catalyzed C-N cross-coupling reactions, α-branched secondary amines are difficult coupling partners and the desired products are often produced in low yields. In order to provide a robust method for accessing N-aryl α-branched tertiary amines, new catalysts have been designed

Articles

G3 and G4 Buchwald palladium precatalysts are the newest air, moisture, and thermally stable crossing-coupling complexes used in bond formation for their versatility and high reactivity.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service