Skip to Content
Merck
All Photos(2)

Key Documents

900456

Sigma-Aldrich

AlkylFluor

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€138.00
1 G
€519.00

€138.00


Estimated to ship on05 April 2025


Request a Bulk Order

Select a Size

Change View
250 MG
€138.00
1 G
€519.00

About This Item

Empirical Formula (Hill Notation):
C27H36BF5N2
CAS Number:
Molecular Weight:
494.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

€138.00


Estimated to ship on05 April 2025


Request a Bulk Order

form

powder

Quality Level

mp

>300 °C

functional group

fluoro

SMILES string

[B-](F)(F)(F)F.Fc1[n+](cc[n]1c3c(cccc3C(C)C)C(C)C)c2c(cccc2C(C)C)C(C)C

InChI key

ZXIKPDSPOFQTLH-UHFFFAOYSA-N

Application

AlkylFluor, a bench-stable salt analogue of PhenoFluor, is a reagent for high-yielding deoxyfluorination of alcohols, including sterically demanding substrates. Insensitive to water and air, AlkylFluor provides convenient access to a broad scope of late-stage fluorinated products as demonstrated by the Ritter Lab.
Suggested reagents to use with AlkylFluor include potassium fluoride (449148) as a fluoride source and 1,4-dioxane (296309) as a solvent.

Legal Information

PhenoFluor is a trademark of SciFluor Life Sciences, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The Ritter lab currently focuses on fluorination chemistry for late-stage functionalization of complex natural and unnatural products. PhenoFluor™ has been developed as a general reagent for the selective, predictable, direct deoxyfluorination of complex alcohols and phenols.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service