900427
SGA 521 100 isiphor
Synonym(s):
Europium doped alkaline earth orthosilicate, Green phosphor, Rare earth doped phosphor
About This Item
form
powder
composition
Halogen free, Br <700 ppm, Cl <700 ppm, Br+Cl <1000 ppm
particle size
12.5-14.5 μm (D50)
20-22 μm (D90)
6-8 μm (D10)
fluorescence
λem 521 nm±1 nm
Application
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service