Skip to Content
Merck
All Photos(1)

Key Documents

900327

Sigma-Aldrich

Poly(D,L-lactide-co-trimethylene carbonate)

lactide:TMC 50:50, viscosity 1.5 dL/g 

Synonym(s):

PDLA-TMC

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€414.00

€414.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€414.00

About This Item

Linear Formula:
CH3(C3H4O2)m(C4H6O3)nCH3
UNSPSC Code:
12162002
NACRES:
NA.23

€414.00


Check Cart for Availability

Request a Bulk Order

Quality Level

form

chips or granules (chunks)

feed ratio

lactide:trimethylene carbonate 50:50

viscosity

1.5 dL/g

storage temp.

−20°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Innovations in polymer technology have had a significant impact on the advancement of novel drug delivery systems.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service