Skip to Content
Merck
All Photos(1)

Key Documents

900277

Sigma-Aldrich

Bis(3,5-bis(trifluoromethyl)phenyl)(2′,6′-bis(dimethylamino)-3,6-dimethoxybiphenyl-2-yl)phosphine

≥95%

Synonym(s):

2′-(Bis(3,5-bis(trifluoromethyl)phenyl)phosphanyl)-3′,6′-dimethoxy-N ,N ,N ,N -tetramethyl-[1,1′-biphenyl]-2,6-diamine, 2′-(Bis(3,5-bis(trifluoromethyl)phenyl)phosphanyl)-3′,6′-dimethoxyN2,N2,N6,N6 -tetramethyl-[1,1′-biphenyl]-2,6-diamine, 2-{Bis[3,5-bis(trifluoromethyl)phenyl]phosphino}-3,6-dimethoxy -2′,6′-bis(dimethylamino)-1,1′-biphenyl

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€495.00
1 G
€1,900.00

€495.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€495.00
1 G
€1,900.00

About This Item

Empirical Formula (Hill Notation):
C34H29F12N2O2P
CAS Number:
Molecular Weight:
756.56
MDL number:
UNSPSC Code:
12352128
NACRES:
NA.22

€495.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

powder or crystals

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

reagent type: ligand

mp

160.3 °C

functional group

phosphine

SMILES string

FC(F)(F)c1cc(cc(c1)C(F)(F)F)P(c3c(ccc(c3c4c(cccc4N(C)C)N(C)C)OC)OC)c2cc(cc(c2)C(F)(F)F)C(F)(F)F

InChI

1S/C34H29F12N2O2P/c1-47(2)24-8-7-9-25(48(3)4)28(24)29-26(49-5)10-11-27(50-6)30(29)51(22-14-18(31(35,36)37)12-19(15-22)32(38,39)40)23-16-20(33(41,42)43)13-21(17-23)34(44,45)46/h7-17H,1-6H3

InChI key

RXBSKAKCDMMHNB-UHFFFAOYSA-N

Application

As the Pd G4 complex, this revolutionary ligand from the Buchwald group offers best-in-class performance in the arylation of sterically hindered secondary amines.

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathaniel H Park et al.
Angewandte Chemie (International ed. in English), 54(28), 8259-8262 (2015-06-03)
In Pd-catalyzed C-N cross-coupling reactions, α-branched secondary amines are difficult coupling partners and the desired products are often produced in low yields. In order to provide a robust method for accessing N-aryl α-branched tertiary amines, new catalysts have been designed

Related Content

The Buchwald group has developed a series of highly active and versatile palladium precatalysts and biarylphosphine ligands used in cross-coupling reactions for the formation of C-C, C–N, C–O, C–F, C–CF3, and C–S bonds. The ligands are electron-rich, and highly tunable to provide catalyst systems with a diverse scope, high stability and reactivity. Furthermore, the new series of precatalysts are air-, moisture and thermally-stable and display good solubility in common organic solvents. The use of precatalysts ensures the efficient generation of the active catalytic species and allows one to accurately adjust the ligand:palladium ratio. The ligands, precatalysts and methodology developed in the Buchwald group are user friendly and have rendered previously difficult cross couplings reactions, much easier to achieve.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service