Skip to Content
Merck
All Photos(1)

Key Documents

900222

Sigma-Aldrich

CdSe/ZnS core-shell type quantum dots

carboxylic acid functionalized, fluorescence λem 580 nm, 1 mg/mL in H2O

Synonym(s):

Fluorescent nanocrystals, QDs

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€156.00

€156.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€156.00

About This Item

Linear Formula:
CdSe/ZnS
UNSPSC Code:
26111700
NACRES:
NA.23

€156.00


Check Cart for Availability

Request a Bulk Order

form

dispersion

Quality Level

concentration

1 mg/mL in H2O

fluorescence

λem 580 nm
λem 580 nm±10 nm, quantum yield >50%

functional group

carboxylic acid

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Reactive group: Carboxylic acid.

Application

The organic layer of the CdS/ZnS or CdSe/ZnS quantum dots (QDs) with carboxylic acid as reactive group consist of a monolayer of oleic acid/octadecylamine and a monolayer of amphiphilic polymer. The total thickness of organic layers on these QDs is approximately 4 nm. They find many applications in bioimaging and biosensing.[1][2][3]

Caution

Do not freeze.

Legal Information

Product of Ocean Nanotech, LLC.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhanao Tan et al.
Nano letters, 7(12), 3803-3807 (2007-11-03)
We report a multilayer solution-processed blue light-emitting diode based on colloidal core/shell CdS/ZnS nanocrystal quantum dots (QDs). At a low-operating voltage of 5.5 V, the device emits spectrally pure blue radiation at 460 nm with a narrow full-width-at-half-maximum bandwidth of
Bright, multicoloured light-emitting diodes based on quantum dots.
Sun Q, et al.
Nature Photonics, 1, 717-722 (2007)
Qingjiang Sun et al.
ACS nano, 3(3), 737-743 (2009-03-25)
Owing to their narrow bright emission band, broad size-tunable emission wavelength, superior photostability, and excellent flexible-substrate compatibility, light-emitting diodes based on quantum dots (QD-LEDs) are currently under intensive research and development for multiple consumer applications including flat-panel displays and flat

Articles

Cadmium-free quantum dots for bioimaging offer design and synthesis strategies, enhancing bioimaging and sensing applications.

Questions

  1. What is the exact size of this QD?

    1 answer
    1. The particle size for this item is 7.4 nm

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service