Skip to Content
Merck
All Photos(1)

Key Documents

88345

Sigma-Aldrich

Thiomorpholine hydrochloride

≥98.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9NS · HCl
CAS Number:
Molecular Weight:
139.65
Beilstein:
3908104
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (AT)

form

crystals

mp

174-177 °C

functional group

thioether

storage temp.

2-8°C

SMILES string

Cl[H].C1CSCCN1

InChI

1S/C4H9NS.ClH/c1-3-6-4-2-5-1;/h5H,1-4H2;1H

InChI key

QSJAHJYXDRUZMY-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J I Levin et al.
Bioorganic & medicinal chemistry letters, 16(6), 1605-1609 (2006-01-24)
A series of thiomorpholine sulfonamide hydroxamate TACE inhibitors, all bearing propargylic ether P1' groups, was explored. In particular, compound 5h has excellent in vitro potency against isolated TACE enzyme and in cells, oral activity in a model of TNF-alpha production
B Combourieu et al.
Biodegradation, 9(6), 433-442 (1999-05-21)
Spectrophotometric assays of Mycobacterium aurum MO1 cells extracts gave evidence of a soluble cytochrome P450, involved in the degradative pathway of morpholine, a waste product from the chemical industry. In order to get further information, the kinetics of the biodegradation
I T Ermakova et al.
Mikrobiologiia, 77(5), 617-622 (2008-11-14)
A screening of lignin-degrading basidial fungi that can grow in the presence of thiomorpholine derivatives (the mixture of 1,4-perhydrothiazines) has been performed. Strain Bjerkandera adusta VKM F-3477 was shown to have the maximal rate of growth in the presence of
B Beck et al.
Molecular diversity, 14(3), 479-491 (2010-04-22)
We designed two novel thiolactone scaffolds. Both scaffolds can be accessed by a convergent Ugi multicomponent reaction (MCR) and are, thus, amenable to library synthesis. Design, stereoselectivity, structures, full experimental details, and virtual libraries will be reported.
Saurav Bera et al.
ACS combinatorial science, 14(1), 1-4 (2011-12-01)
Diastereoselective trans-2,5-disubstituted amino acids derived diverse morpholines, piperazines and thiomorpholines were prepared in 30 min-1 h with high yields through iodine-mediated 6-exotrig type cyclization from a single common synthetic intermediate. The displacement of iodine with hydride ion gave a methyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service