Skip to Content
Merck
All Photos(1)

Key Documents

87600

Sigma-Aldrich

N,N,N′,N′-Tetrakis(2-hydroxyethyl)ethylenediamine

technical

Synonym(s):

2,2′,2″,2″′-(Ethylenedinitrilo)tetraethanol, THEED

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€40.80

€40.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€40.80

About This Item

Linear Formula:
(HOCH2CH2)2NCH2CH2N(CH2CH2OH)2
CAS Number:
Molecular Weight:
236.31
Beilstein:
1767829
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

€40.80


Check Cart for Availability

Request a Bulk Order

grade

technical

form

liquid

refractive index

n20/D 1.501

bp

~280 °C

density

1.1 g/mL at 20 °C (lit.)

functional group

amine
hydroxyl

SMILES string

OCCN(CCO)CCN(CCO)CCO

InChI

1S/C10H24N2O4/c13-7-3-11(4-8-14)1-2-12(5-9-15)6-10-16/h13-16H,1-10H2

InChI key

BYACHAOCSIPLCM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N,N′,N′-Tetrakis(2-hydroxyethyl)ethylenediamine, is primarily used as a chelating agent and as a building block in the synthesis of various chemical compounds. It is also useful in metal extraction and catalysis because of its ability to form stable complexes with metal ions.

Application

N,N,N′,N′-Tetrakis(2-hydroxyethyl)ethylenediamine can be used as a chelating agent to prepare a palladium-based complex, which is used as a catalyst for the Suzuki/Miyaura reaction.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marko Pende et al.
Science advances, 6(22), eaba0365-eaba0365 (2020-06-12)
Tissue clearing combined with deep imaging has emerged as a powerful alternative to classical histological techniques. Whereas current techniques have been optimized for imaging selected nonpigmented organs such as the mammalian brain, natural pigmentation remains challenging for most other biological

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service