862177
α-D-Glucose 1-phosphate disodium salt hydrate
98%
Synonym(s):
α-D-Glucopyranose 1-phosphate, Cori ester
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H11O9PNa2 · xH2O
CAS Number:
Molecular Weight:
304.10 (anhydrous basis)
MDL number:
UNSPSC Code:
12352201
Assay
98%
optical activity
[α]21/D +79.9°, c = 4.1 in H2O
SMILES string
[Na+].[Na+].[H]O[H].OC[C@H]1O[C@H](OP([O-])([O-])=O)[C@H](O)[C@@H](O)[C@@H]1O
InChI
1S/C6H13O9P.2Na.H2O/c7-1-2-3(8)4(9)5(10)6(14-2)15-16(11,12)13;;;/h2-10H,1H2,(H2,11,12,13);;;1H2/q;2*+1;/p-2/t2-,3-,4+,5-,6-;;;/m1.../s1
InChI key
YWAUQXHOCBBYLP-FBNUBEQJSA-L
Looking for similar products? Visit Product Comparison Guide
General description
α-D-Glucose 1-phosphate is converted into D-glucose-6-phosphate by the enzyme phosphoglucomutase.
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Joerg Fettke et al.
Plant physiology, 155(4), 1723-1734 (2010-12-01)
Almost all glucosyl transfer reactions rely on glucose-1-phosphate (Glc-1-P) that either immediately acts as glucosyl donor or as substrate for the synthesis of the more widely used Glc dinucleotides, ADPglucose or UDPglucose. In this communication, we have analyzed two Glc-1-P-related
Kyra-Melinda Alexacou et al.
Bioorganic & medicinal chemistry, 18(22), 7911-7922 (2010-10-16)
Glycogen phosphorylase (GP) is a promising target for the treatment of type 2 diabetes. In the process of structure based drug design for GP, a group of 15 aromatic aldehyde 4-(β-d-glucopyranosyl)thiosemicarbazones have been synthesized and evaluated as inhibitors of rabbit
Bijay Singh et al.
Protein engineering, design & selection : PEDS, 25(4), 179-187 (2012-02-16)
Two similar genes, dnmL and rmbA in Streptomyces peucetius, which encode for glucose-1-phosphate (G-1-P) thymidylyltransferases were expressed in Escherichia coli under similar conditions. While RmbA was expressed in soluble form, DnmL was found as insoluble aggregates in inclusion bodies. The
Jef Van der Borght et al.
Biotechnology journal, 5(9), 986-993 (2010-08-28)
β-D-Glucose-1-phosphate (βGlc1P) is an efficient glucosyl donor for both enzymatic and chemical glycosylation reactions but is currently very costly and not available in large amounts. This article provides an efficient production method of βGlc1P from trehalose and phosphate using the
Stanley A Blumenthal
Perspectives in biology and medicine, 55(2), 236-249 (2012-05-31)
In 1945, Earl Sutherland (1915-1974) [corrected] and associates began studies of the mechanism of hormone-induced glycogen breakdown in the liver. In 1956, their efforts culminated in the identification of cyclic AMP, an ancient molecule generated in many cell types in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service