Skip to Content
Merck
All Photos(1)

Key Documents

861863

Sigma-Aldrich

2-Chloroadenosine hemihydrate

97%

Synonym(s):

6-Amino-2-chloropurine riboside

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€134.00

€134.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€134.00

About This Item

Empirical Formula (Hill Notation):
C10H12ClN5O4 · 0.5H2O
CAS Number:
Molecular Weight:
310.69
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€134.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

powder

mp

160 °C (dec.) (lit.)

functional group

chloro
ether

SMILES string

O[C@H]1[C@@H](O)[C@H](N(C=N2)C3=C2C(N)=NC(Cl)=N3)O[C@@H]1CO.O[C@H]4[C@@H](O)[C@H](N(C=N5)C6=C5C(N)=NC(Cl)=N6)O[C@@H]4CO.O

InChI

1S/2C10H12ClN5O4.H2O/c2*11-10-14-7(12)4-8(15-10)16(2-13-4)9-6(19)5(18)3(1-17)20-9;/h2*2-3,5-6,9,17-19H,1H2,(H2,12,14,15);1H2/t2*3-,5-,6-,9-;/m11./s1

InChI key

WRURCFOLSGPTMO-IZGCVNAISA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian H Trieu et al.
The Journal of physiology, 593(13), 2889-2907 (2015-04-24)
Extended trains of theta rhythm afferent activity lead to a biphasic response facilitation in field CA1 but not in the lateral perforant path input to the dentate gyrus. Processes that reverse long-term potentiation in field CA1 are not operative in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service