Skip to Content
Merck
All Photos(1)

Key Documents

860956

Sigma-Aldrich

Congo Red

certified by the Biological Stain Commission, Dye content 85 %

Synonym(s):

Congo Red 4B, Cosmos Red, Cotton Red B, Cotton Red C, Direct Red 28, Direct Red R, Direct Red Y

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H22N6Na2O6S2
CAS Number:
Molecular Weight:
696.66
Colour Index Number:
22120
Beilstein:
3894858
EC Number:
MDL number:
UNSPSC Code:
12171500

grade

certified by the Biological Stain Commission

composition

Dye content, 85%

technique(s)

titration: suitable

pH

3.0-5.0, blue to red

mp

>360 °C (lit.)

λmax

497 nm

SMILES string

[Na+].[Na+].Nc1c(cc(c2ccccc12)S([O-])(=O)=O)\N=N\c3ccc(cc3)-c4ccc(cc4)\N=N\c5cc(c6ccccc6c5N)S([O-])(=O)=O

InChI

1S/C32H24N6O6S2.2Na/c33-31-25-7-3-1-5-23(25)29(45(39,40)41)17-27(31)37-35-21-13-9-19(10-14-21)20-11-15-22(16-12-20)36-38-28-18-30(46(42,43)44)24-6-2-4-8-26(24)32(28)34;;/h1-18H,33-34H2,(H,39,40,41)(H,42,43,44);;/q;2*+1/p-2/b37-35+,38-36+;;

InChI key

IQFVPQOLBLOTPF-HKXUKFGYSA-L

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jean D Sipe et al.
Amyloid : the international journal of experimental and clinical investigation : the official journal of the International Society of Amyloidosis, 19(4), 167-170 (2012-11-02)
The Nomenclature Committee of the International Society of Amyloidosis (ISA) met during the XIIIth International Symposium, May 6-10, 2012, Groningen, The Netherlands, to formulate recommendations on amyloid fibril protein nomenclature and to consider newly identified candidate amyloid fibril proteins for
A Yu Mironenko et al.
Carbohydrate polymers, 92(1), 769-774 (2012-12-12)
Sensitive layer-by-layer (LbL) coatings for optical detection of gaseous NH(3) and HCl were prepared by self-assembly of oppositely charged polysaccharides (chitosan and λ-carrageenan) followed by doping LbLs with pH-sensitive dyes - bromothymol blue (BTB) and Congo red (CR). It has
Guangshu Zhang et al.
Dalton transactions (Cambridge, England : 2003), 42(10), 3660-3666 (2013-01-10)
Novel nickel hierarchical structures were prepared through a facile one-pot solvothermal approach in the presence of surfactant and mineralizer. The influence of dynamics factors such as polyvinylpyrrolidone (PVP) content, ratio (R) between NaF and NH(3)·H(2)O on the morphologies of the
Cong Liu et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(51), 20913-20918 (2012-12-06)
Although aberrant protein aggregation has been conclusively linked to dozens of devastating amyloid diseases, scientists remain puzzled about the molecular features that render amyloid fibrils or small oligomers toxic. Here, we report a previously unobserved type of amyloid fibril that
Rita A Rapa et al.
PloS one, 8(3), e58430-e58430 (2013-03-14)
The integron is a genetic recombination system that catalyses the acquisition of genes on mobilisable elements called gene cassettes. In Vibrio species, multiple acquired gene cassettes form a cassette array that can comprise 1-3% of the bacterial genome. Since 75%

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service