Skip to Content
Merck
All Photos(1)

Key Documents

859990

Sigma-Aldrich

2-Acetamido-2-deoxy-β-D-glucopyranose 1,3,4,6-tetraacetate

98%

Synonym(s):

β-D-Glucosamine pentaacetate, 2-Amino-2-deoxy-β-D-glucopyranosyl pentaacetate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€271.00

€271.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€271.00

About This Item

Empirical Formula (Hill Notation):
C16H23NO10
CAS Number:
Molecular Weight:
389.35
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

€271.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

optical activity

[α]24/D +1.7 to +3.0°, c = 10 in chloroform

mp

186-189 °C (lit.)

storage temp.

−20°C

SMILES string

CC(=O)N[C@H]1[C@@H](O[C@H](COC(C)=O)[C@@H](OC(C)=O)[C@@H]1OC(C)=O)OC(C)=O

InChI

1S/C16H23NO10/c1-7(18)17-13-15(25-10(4)21)14(24-9(3)20)12(6-23-8(2)19)27-16(13)26-11(5)22/h12-16H,6H2,1-5H3,(H,17,18)/t12-,13-,14-,15-,16-/m1/s1

InChI key

OVPIZHVSWNOZMN-OXGONZEZSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alicja K Antonczak et al.
The Journal of biological chemistry, 284(42), 28795-28800 (2009-08-12)
The ability of Escherichia coli to grow on a series of acetylated and glycosylated compounds has been investigated. It is surmised that E. coli maintains low levels of nonspecific esterase activity. This observation may have ramifications for previous reports that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service