Skip to Content
Merck
All Photos(1)

Documents

856576

Sigma-Aldrich

Adogen® 464

Synonym(s):

Methyltrialkyl(C8-C10)ammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

>1 (vs air)

vapor pressure

33 mmHg ( 20 °C)

form

liquid

expl. lim.

12 %

refractive index

n20/D 1.4665 (lit.)

density

0.898 g/mL at 25 °C (lit.)

SMILES string

[Cl-].[Cl-].[Cl-].CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC.CCCCCCCCC[N+](C)(CCCCCCCCC)CCCCCCCCC.CCCCCCCCCC[N+](C)(CCCCCCCCCC)CCCCCCCCCC

InChI

1S/C31H66N.C28H60N.C25H54N.3ClH/c1-5-8-11-14-17-20-23-26-29-32(4,30-27-24-21-18-15-12-9-6-2)31-28-25-22-19-16-13-10-7-3;1-5-8-11-14-17-20-23-26-29(4,27-24-21-18-15-12-9-6-2)28-25-22-19-16-13-10-7-3;1-5-8-11-14-17-20-23-26(4,24-21-18-15-12-9-6-2)25-22-19-16-13-10-7-3;;;/h5-31H2,1-4H3;5-28H2,1-4H3;5-25H2,1-4H3;3*1H/q3*+1;;;/p-3

InChI key

RMQKQYWUFZNCMA-UHFFFAOYSA-K

Application

Common phase transfer catalyst. Used to extract metal ions for spectrophotometric determinations.

Legal Information

Adogen is a registered trademark of Evonik Goldschmidt Corporation

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 3 - Repr. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

109.9 °F - closed cup

Flash Point(C)

43.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fresenius Journal of Analytical Chemistry, 344, 30-30 (1992)
Rev. Roum. Chim., 36, 77-77 (1991)
Dehmlow, E.V. Dehmlow, S.S.
Phase Transfer Catalysis (Academic Press) (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service