Skip to Content
Merck
All Photos(1)

Key Documents

856541

Sigma-Aldrich

Sodium 4-aminosalicylate dihydrate

99%

Synonym(s):

4-Amino-2-hydroxybenzoic acid sodium salt, 4-Amino-salicylic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6NNaO3 · 2H2O
CAS Number:
Molecular Weight:
211.15
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

99%

SMILES string

O.O.[Na+].Nc1ccc(c(O)c1)C([O-])=O

InChI

1S/C7H7NO3.Na.2H2O/c8-4-1-2-5(7(10)11)6(9)3-4;;;/h1-3,9H,8H2,(H,10,11);;2*1H2/q;+1;;/p-1

InChI key

GMUQJDAYXZXBOT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sumit Chakraborty et al.
Science (New York, N.Y.), 339(6115), 88-91 (2012-11-03)
Folate biosynthesis is an established anti-infective target, and the antifolate para-aminosalicylic acid (PAS) was one of the first anti-infectives introduced into clinical practice on the basis of target-based drug discovery. Fifty years later, PAS continues to be used to treat
Cezary Chojnacki et al.
Polski merkuriusz lekarski : organ Polskiego Towarzystwa Lekarskiego, 31(182), 92-96 (2011-09-23)
In order to maintain ulcerative colitis (UC) in remission, chronic use of aminosalicylates is recommended. All patients have a fear of the recurrence of symptoms, which makes their mental state and quality of life worse. Because of this a number
A C Liwa et al.
Journal of tropical pediatrics, 59(2), 90-94 (2012-11-24)
There are no paediatric data regarding slow-release para-aminosalicylic acid (PAS). We studied PAS plasma concentrations in 10 children receiving a single 150 mg/kg dose daily or 75 mg/kg twice daily and 12 adults receiving 4 g twice daily. Blood specimens
Amanda S Fivian-Hughes et al.
Microbiology (Reading, England), 158(Pt 2), 308-318 (2011-10-29)
Thymidylate synthase (TS) enzymes catalyse the biosynthesis of deoxythymidine monophosphate (dTMP or thymidylate), and so are important for DNA replication and repair. Two different types of TS proteins have been described (ThyA and ThyX), which have different enzymic mechanisms and
S M Vesenbeckh et al.
Infection, 40(2), 199-202 (2011-08-13)
para-Aminosalicylic acid (PAS) is commonly used in the treatment of drug-resistant tuberculosis, including multidrug-resistant tuberculosis. Since its first use in the 1940s, hypersensitivity reactions frequently limit its use in clinical practice. Cases of successful desensitization against PAS using orally administered

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service