Skip to Content
Merck
All Photos(1)

Key Documents

856495

Sigma-Aldrich

Potassium 4-nitrophenyl sulfate

≥99%

Synonym(s):

4-Nitrophenyl sulfate potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NO2C6H4OSO2OK
CAS Number:
Molecular Weight:
257.26
Beilstein:
3786392
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

≥99%

mp

246-250 °C (lit.)

SMILES string

[K+].[O-][N+](=O)c1ccc(OS([O-])(=O)=O)cc1

InChI

1S/C6H5NO6S.K/c8-7(9)5-1-3-6(4-2-5)13-14(10,11)12;/h1-4H,(H,10,11,12);/q;+1/p-1

InChI key

BITVAZYUWRLLCN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service