Skip to Content
Merck
All Photos(2)

Key Documents

855987

Sigma-Aldrich

D-Valine

≥98%, for peptide synthesis

Synonym(s):

(R)-α-Aminoisovaleric acid, D-2-Amino-3-methylbutanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€51.50
25 G
€105.00

€51.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€51.50
25 G
€105.00

About This Item

Linear Formula:
(CH3)2CHCH(NH2)CO2H
CAS Number:
Molecular Weight:
117.15
Beilstein:
1721135
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

€51.50


Check Cart for Availability

Request a Bulk Order

Product Name

D-Valine, ≥98%

Quality Level

Assay

≥98%

form

powder

optical activity

[α]23/D −32.0 to −24.0°, c = 8 in 6 M HCl

optical purity

ee: 99% (GLC)

reaction suitability

reaction type: solution phase peptide synthesis

mp

>295 °C (subl.) (lit.)

application(s)

peptide synthesis

SMILES string

CC(C)[C@@H](N)C(O)=O

InChI

1S/C5H11NO2/c1-3(2)4(6)5(7)8/h3-4H,6H2,1-2H3,(H,7,8)/t4-/m1/s1

InChI key

KZSNJWFQEVHDMF-SCSAIBSYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Kimberly A Smith et al.
American journal of respiratory cell and molecular biology, 53(3), 355-367 (2015-01-09)
Hypoxic pulmonary vasoconstriction (HPV) is an important physiological response that optimizes the ventilation/perfusion ratio. Chronic hypoxia causes vascular remodeling, which is central to the pathogenesis of hypoxia-induced pulmonary hypertension (HPH). We have previously shown that Notch3 is up-regulated in HPH
Shanshan Song et al.
American journal of physiology. Cell physiology, 307(4), C373-C383 (2014-06-13)
An increase in cytosolic Ca(2+) concentration ([Ca(2+)]cyt) in pulmonary arterial smooth muscle cells (PASMC) is a major trigger for pulmonary vasoconstriction and an important stimulus for pulmonary arterial medial hypertrophy in patients with idiopathic pulmonary arterial hypertension (IPAH). Vascular smooth
Stefan M Gysler et al.
American journal of reproductive immunology (New York, N.Y. : 1989), 73(3), 242-250 (2014-07-30)
Women with antiphospholipid syndrome (APS) are at increased risk of recurrent pregnancy loss (RPL) and preeclampsia. Antiphospholipid antibodies (aPL) directly alter trophoblast function. Treatment with low molecular weight heparin (LMWH) reduces the risk of RPL but not preeclampsia. Moreover, LMWH
Dean P J Kavanagh et al.
Stem cells (Dayton, Ohio), 33(9), 2785-2797 (2015-07-01)
Mesenchymal stem cells (MSCs) have shown therapeutic promise in many experimental and clinical models of inflammation. However, a commonly reported feature of MSC transplantation is poor homing to injured tissues. Previously, we have shown that pretreatment with cytokines/chemical factors enhances
Gerald I Richard et al.
Applied spectroscopy, 62(5), 476-480 (2008-05-24)
The spectroscopic properties of a chiral boronic acid based resorcinarene macrocycle employed for chiral analysis were investigated. Specifically, the emission and excitation characteristics of tetraarylboronate resorcinarene macrocycle (TBRM) and its quantum yield were evaluated. The chiral selector TBRM was investigated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service