Skip to Content
Merck
All Photos(1)

Key Documents

852430

Sigma-Aldrich

N6-Benzyladenine

99%

Synonym(s):

6-Benzylaminopurine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11N5
CAS Number:
Molecular Weight:
225.25
Beilstein:
19406
EC Number:
MDL number:
UNSPSC Code:
12352100

Assay

99%

mp

230-233 °C (lit.)

SMILES string

C(Nc1ncnc2nc[nH]c12)c3ccccc3

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

You-Hua Wang et al.
Ying yong sheng tai xue bao = The journal of applied ecology, 22(5), 1233-1239 (2011-08-05)
A field trial with high fiher quality cotton cultivar Kemian 1 was conducted in Nanjing (lower reaches of Yangtze River) in 2006-2007 to study the effects of growth regulators 6-BA and ABA on the boll and fiber development and related
E S Davidiants
Prikladnaia biokhimiia i mikrobiologiia, 47(5), 530-536 (2012-01-12)
Influence of the aleanolic acid glycosides from Silphium perfoliatum L. (silphioside B, C, E and G) and their progenins on the amylase activity and total protein content in wheat seedlings was studied. Treatment of the Triticum aestivum L. seeds with
Priyanka Srivastava et al.
Plant signaling & behavior, 6(7), 974-981 (2011-06-28)
Present report is the first direct evidence of azadirachtin production in androgenic haploid cultures of Azadirachta indica, a woody medicinal tree. Anther cultures at early-late-uninucleate stage of microspores were established on MS medium with BAP (5 μM), 2,4-D (1 μM)
Nils Braun et al.
Plant physiology, 158(1), 225-238 (2011-11-03)
The function of PsBRC1, the pea (Pisum sativum) homolog of the maize (Zea mays) TEOSINTE BRANCHED1 and the Arabidopsis (Arabidopsis thaliana) BRANCHED1 (AtBRC1) genes, was investigated. The pea Psbrc1 mutant displays an increased shoot-branching phenotype, is able to synthesize strigolactone
V Kalita et al.
Cryo letters, 33(1), 58-68 (2012-03-22)
The present investigation was aimed at developing a protocol for long-term preservation of germplasm of Pinus kesiya Royle ex. Gord. through vitrification. Some of the critical components affecting explant tolerance to cryopreservation, such as effects of preculture, vitrification solutions, exposure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service