Skip to Content
Merck
All Photos(1)

Key Documents

852279

Sigma-Aldrich

2′-Deoxyinosine

99%

Synonym(s):

9-(2-Deoxy-β-D-ribofuranosyl)hypoxanthine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N4O4
CAS Number:
Molecular Weight:
252.23
Beilstein:
33517
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Pricing and availability is not currently available.

Quality Level

Assay

99%

form

powder

functional group

ether
hydroxyl

SMILES string

OC[C@H]1O[C@H](C[C@@H]1O)n2cnc3C(=O)NC=Nc23

InChI

1S/C10H12N4O4/c15-2-6-5(16)1-7(18-6)14-4-13-8-9(14)11-3-12-10(8)17/h3-7,15-16H,1-2H2,(H,11,12,17)/t5-,6+,7+/m0/s1

InChI key

VGONTNSXDCQUGY-RRKCRQDMSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manabu Yasui et al.
Journal of molecular biology, 377(4), 1015-1023 (2008-02-29)
Chronic inflammation involving constant generation of nitric oxide (*NO) by macrophages has been recognized as a factor related to carcinogenesis. At the site of inflammation, nitrosatively deaminated DNA adducts such as 2'-deoxyinosine (dI) and 2'-deoxyxanthosine are primarily formed by *NO
Bernard Weiss
DNA repair, 7(2), 205-212 (2007-11-06)
Deoxyinosine (dI) is produced in DNA by the hydrolytic or nitrosative deamination of deoxyadenosine. It is excised in a repair pathway that is initiated by endonuclease V, the product of the nfi gene. The repair was studied in vivo using
Rongjuan Mi et al.
Mutation research, 735(1-2), 12-18 (2012-06-06)
The human endonuclease V gene is located in chromosome 17q25.3 and encodes a 282 amino acid protein that shares about 30% sequence identity with bacterial endonuclease V. This study reports biochemical properties of human endonuclease V with respect to repair
A Cohen et al.
The New England journal of medicine, 295(26), 1449-1454 (1976-12-23)
To delineate the normal function of purine nucleoside phosphorylase and to understand the pathogenesis of the immune dysfunction associated with deficiency of this enzyme, we studied purine metabolism in a patient deficient in purine nucleoside phosphorylase, her erythrocytes and cultured
Norman E Watkins et al.
Nucleic acids research, 33(19), 6258-6267 (2005-11-03)
Nearest-neighbor thermodynamic parameters of the 'universal pairing base' deoxyinosine were determined for the pairs I.C, I.A, I.T, I.G and I.I adjacent to G.C and A.T pairs. Ultraviolet absorbance melting curves were measured and non-linear regression performed on 84 oligonucleotide duplexes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service