Skip to Content
Merck
All Photos(1)

Key Documents

851620

Sigma-Aldrich

D-Norvaline

99%

Synonym(s):

(R)-(−)-2-Aminopentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
117.15
Beilstein:
1721161
EC Number:
MDL number:
UNSPSC Code:
12352200
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

crystals

optical activity

[α]20/D −24°, c = 10 in 5 M HCl

optical purity

ee: 98% (GLC)

reaction suitability

reaction type: solution phase peptide synthesis

mp

>300 °C (lit.)

application(s)

peptide synthesis

SMILES string

CCC[C@@H](N)C(O)=O

InChI

1S/C5H11NO2/c1-2-3-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)/t4-/m1/s1

InChI key

SNDPXSYFESPGGJ-SCSAIBSYSA-N

Physical form

α-Amino acid analog with unnatural, (R)-configuration

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Xiaofang Tang et al.
Kidney & blood pressure research, 44(2), 245-263 (2019-05-10)
Diabetic nephropathy (DN) is a leading cause of end-stage renal disease. Microalbuminuria (MA) is widely used to predict early progressive renal function decline (ERFD) of DN in type 2 diabetes mellitus (T2D) patients, but the sensitivity and specificity of MA
Arina G Shokhina et al.
Redox biology, 21, 101071-101071 (2018-12-24)
Redox-sensitive fluorescent proteins (roFPs) are a powerful tool for imaging intracellular redox changes. The structure of these proteins contains a pair of cysteines capable of forming a disulfide upon oxidation that affects the protein conformation and spectral characteristics. To date
Iris Thondorf et al.
Bioorganic & medicinal chemistry, 19(21), 6409-6418 (2011-10-01)
The proton-coupled amino acid transporter hPAT1 has recently gained much interest due to its ability to transport small drugs thereby allowing their oral administration. A three-dimensional quantitative structure-activity relationship (3D QSAR) study has been performed on its natural and synthetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service