Skip to Content
Merck
All Photos(1)

Key Documents

84320

Sigma-Aldrich

Menthyl salicylate

mixture of isomers

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€87.20

€87.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€87.20

About This Item

Empirical Formula (Hill Notation):
C17H24O3
CAS Number:
Molecular Weight:
276.37
Beilstein:
3097754
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€87.20


Check Cart for Availability

Request a Bulk Order

form

liquid

refractive index

n20/D 1.52

bp

186-190 °C/12 mmHg (lit.)

density

1.04 g/mL at 20 °C (lit.)

functional group

ester

SMILES string

CC(C)C1CCC(C)CC1OC(=O)c2ccccc2O

InChI

1S/C17H24O3/c1-11(2)13-9-8-12(3)10-16(13)20-17(19)14-6-4-5-7-15(14)18/h4-7,11-13,16,18H,8-10H2,1-3H3

InChI key

SJOXEWUZWQYCGL-UHFFFAOYSA-N

Application

light stabilizer

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service