Skip to Content
Merck
All Photos(1)

Key Documents

83885

Sigma-Aldrich

Rink amide (aminomethyl)polystyrene

extent of labeling: ~1.1 mmol/g loading

Synonym(s):

4-[(2,4-Dimethoxyphenyl)(Fmoc-amino)methyl]phenoxyacetomido methyl resin, Fmoc Rink amide, Knorr Linker amide resin

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€192.00
5 G
€415.00

€192.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€192.00
5 G
€415.00

About This Item

MDL number:
UNSPSC Code:
12352116
NACRES:
NA.22

€192.00


Check Cart for Availability

Request a Bulk Order

form

solid

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

extent of labeling

~1.1 mmol/g loading

matrix

crosslinked with 1% DVB

particle size

100-200 mesh

application(s)

peptide synthesis

functional group

amine

Looking for similar products? Visit Product Comparison Guide

Application

  • Solid support for Fmoc-based solid phase peptide synthesis (SPPS) of peptide amides.
  • This solid support contains a Rink amide linker attached to aminomethylpolystyrene resin.
  • Cleavage of peptides is achieved by adding 95% TFA.

Other Notes

Review[1][2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tetrahedron, 53, 5643-5643 (1997)
P.H.H. Hermkens et al.
Tetrahedron, 52, 4527-4527 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service