Skip to Content
Merck
All Photos(1)

Key Documents

83711

Sigma-Aldrich

Rhodium on activated charcoal

5% rhodium (Rh) basis

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€399.00

€399.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€399.00

About This Item

UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

€399.00


Check Cart for Availability

Request a Bulk Order

form

solid

reaction suitability

core: rhodium
reagent type: catalyst

concentration

5% Rh

Storage Class Code

11 - Combustible Solids

WGK

nwg

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service