Skip to Content
Merck
All Photos(1)

Key Documents

809659

Sigma-Aldrich

Pyridoxine-(methyl-d3) hydrochloride

≥98 atom % D, ≥96% (CP)

Synonym(s):

Vitamin B6 (pyridoxine-(methyl-d3) hydrochloride)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€387.00

€387.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 MG
€387.00

About This Item

Empirical Formula (Hill Notation):
C8D3H8NO3·HCl
CAS Number:
Molecular Weight:
208.66
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

€387.00


Check Cart for Availability

Request more information

isotopic purity

≥98 atom % D

Quality Level

Assay

≥96% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

color

white to off-white

mass shift

M+3

storage temp.

−20°C

SMILES string

OC1=C(C([2H])([2H])[2H])N=CC(CO)=C1CO.Cl

InChI

1S/C8H11NO3.ClH/c1-5-8(12)7(4-11)6(3-10)2-9-5;/h2,10-12H,3-4H2,1H3;1H/i1D3;

InChI key

ZUFQODAHGAHPFQ-NIIDSAIPSA-N

Related Categories

General description

Pyridoxine hydrochloride is HCl salt of pyridoxine or vitamin B6. It is a water-soluble vitamin which plays an important role in the treatment of roughness, sunburn, acne, and itch of the skin.[1][2]
Pyridoxine-(methyl-d3) hydrochloride is a deuterated vitamin B6 wherein C-2 methyl protons are replaced by deuterium.

Application

Pyridoxine-(methyl-d3) hydrochloride can be used as a stable isotope internal standard for accurate data interpretation in specific biological samples.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical behaviour of pyridoxine hydrochloride (vitamin B6) at carbon paste electrode modified with crown ethers
Desai PB, et al.
Journal of Solid State Electrochemistry, 12(9), 1067-1075 (2008)
Spectral diagnostics of the interaction between pyridoxine hydrochloride and bovine serum albumin in vitro
Zhang H, et al.
Molecular Biology Reports, 35(4), 699-705 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service