Skip to Content
Merck
All Photos(2)

Key Documents

809489

Sigma-Aldrich

Poly(N-isopropyl acrylamide-co-(N-methacryloxysuccinimide))

NHS 10 mol%, average Mn 5,000

Synonym(s):

PNIPAM

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C4H7O2[C6H11NO]x[C8H9NO3]yC13H25S3
UNSPSC Code:
12352200

form

powder

mol wt

average Mn 5,000

storage temp.

2-8°C

Application

This copolymer is 90 mol% N-isopropylacrylamide, 10 mol% NHS activated ester. The NHS ester allows conjugation to a variety of functional groups. The end group functionality contains a carboxylic acid on one end.

Other Notes

Sold for research purposes only: See sigma-aldrich.com/raftlicense Patents: WO98/01478, WO99/311444.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Professor Mitsuhiro Ebara provides insights on several types of smart nanofiber mesh systems that have been explored for different drug delivery purposes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service