Skip to Content
Merck
All Photos(1)

Key Documents

808512

Sigma-Aldrich

γ-[(Propargyl)-imido]-ATP sodium salt

Synonym(s):

Adenosine-5’-[γ-(propargyl)-imido]triphosphate sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€248.00

€248.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€248.00

About This Item

Empirical Formula (Hill Notation):
C13H19N6O12P3 · xNa+
Molecular Weight:
544.24 (free acid basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

€248.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95% (HPLC)

Quality Level

form

solid

reaction suitability

reaction type: click chemistry

solubility

10 mM Tris-HCl, pH 7.5: soluble

shipped in

wet ice

storage temp.

−20°C

SMILES string

NC1=NC=NC2=C1N=CN2[C@H]3O[C@H](COP(OP(O)(OP(O)(NCC#C)=O)=O)(O)=O)[C@@H](O)[C@H]3O

InChI

1S/C13H19N6O12P3/c1-2-3-18-32(22,23)30-34(26,27)31-33(24,25)28-4-7-9(20)10(21)13(29-7)19-6-17-8-11(14)15-5-16-12(8)19/h1,5-7,9-10,13,20-21H,3-4H2,(H,24,25)(H,26,27)(H2,14,15,16)(H2,18,22,23)/t7-,9-,10-,13?/m1/s1

InChI key

PIPOEUYSJJTELD-RJNFYWFKSA-N

Application

Lee, et al,.[1] reported a non-radioactive version of in vitro phosphorylation where γ-[(Propargyl)-imido]-ATP (compound 1)[1] was successfully used instead of γ-32P-modified ATP to phosphorylate GST-tagged recombinant p27kip1 with protein kinase cdk2. The phosphorylated, alkyne-modified protein substrate can be labeled with azides of biotin or fluorescent dyes using Cu(I)-catalyzed click chemistry.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah E Lee et al.
Bioorganic & medicinal chemistry letters, 19(14), 3804-3807 (2009-05-05)
We hereby present a simple yet novel chemical synthesis of a family of gamma-modified ATPs bearing functional groups on the gamma-phosphate that are amenable to further derivatization by highly selective chemical manipulations (e.g., click chemistry, Staudinger ligations). A preliminary screen

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service