Skip to Content
Merck
All Photos(3)

Key Documents

807184

Sigma-Aldrich

Silver nanospheres

80 nm avg. part. size, 1 mg/mL in water, PVP functionalized

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€388.00

€388.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€388.00

About This Item

UNSPSC Code:
12141740
NACRES:
NA.23

€388.00


Check Cart for Availability

Request a Bulk Order

form

nanospheres

Quality Level

concentration

1 mg/mL in water

avg. part. size

80 nm

λmax

455 nm

functional group

PVP

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Silver nanomaterials have unique physical, chemical and optical properties that are currently being leveraged for a wide variety of biological applications. Additionally, silver nanoparticles have an optical color that is a function of the silver nanoparticle′s size and shape. The strong coupling of silver nanoparticles to specific wavelengths of incident light can be utilized to develop ultrabright reporter molecules, highly efficient thermal absorbers, and nanoscale “antennas” that can amplify the strength of the local electromagnetic field.

Preparation Note

Shake well before use.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service