Skip to Content
Merck
All Photos(1)

Key Documents

80613

Sigma-Aldrich

(−)-trans-Pinocarveol

≥96.0% (sum of enantiomers, GC)

Synonym(s):

(1S,3R,5S)-2(10)-Pinen-3-ol, (1S,3R,5S)-6,6-Dimethyl-2-methylenebicyclo[3.1.1]heptan-3-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Beilstein:
5730571
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96.0% (sum of enantiomers, GC)

form

liquid

optical activity

[α]20/D −72±3°, neat

color

opaque white

refractive index

n20/D 1.500

density

0.979 g/mL at 20 °C (lit.)

SMILES string

CC1(C)[C@@H]2C[C@@H](O)C(=C)[C@H]1C2

InChI

1S/C10H16O/c1-6-8-4-7(5-9(6)11)10(8,2)3/h7-9,11H,1,4-5H2,2-3H3/t7-,8+,9+/m0/s1

InChI key

LCYXQUJDODZYIJ-DJLDLDEBSA-N

General description

(-)-trans-Pinocarveol is an allyl alcohol. It is formed during the allylic oxidation of β-pinene using hydrogen peroxide-selenium dioxide or from the isomerization of α-pinene oxide over zirconium oxide.

Other Notes

Volatile component of essential oils occurring in many plants. Starting material for highly pure (-)-nopinone

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isomerization of a-pinene oxide over solid acids and bases.
Arata K and Tanabe K.
Chemistry Letters (Jpn), 8(8), 1017-1018 (1979)
Allylic Oxidation with Hydrogen Peroxide-Selenium Dioxide: trans-Pinocarveol.
Coxon JM, et al.
Organic Syntheses, 25-25 (1977)
P. Lavalee et al.
The Journal of Organic Chemistry, 51, 1362-1362 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service