Skip to Content
Merck
All Photos(2)

Key Documents

804320

Sigma-Aldrich

Potassium 4-ethoxycarbonylbenzoyltrifluoroborate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€127.00

€127.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€127.00

About This Item

Empirical Formula (Hill Notation):
C10H9BF3KO3
CAS Number:
Molecular Weight:
284.08
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

€127.00


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

mp

228.00 °C

SMILES string

O=C(B(F)(F)F)C1=CC=C(C(OCC)=O)C=C1.[K]

InChI

1S/C10H9BF3O3.K.H/c1-2-17-10(16)8-5-3-7(4-6-8)9(15)11(12,13)14;;/h3-6H,2H2,1H3;;

InChI key

YXYMQBSRJGUKGK-UHFFFAOYSA-N

Application

Acyltrifluroborates (KATs) are bench, air, and moisture stable reagents for rapid, chemoselective amide bond formations with hydroxylamines. These amide bond forming reactions proceed under aqueous conditions, without the need for coupling reagents or protecting groups. This product was introduced in collaboration with the Bode Research Group

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service