Skip to Content
Merck
All Photos(1)

Documents

803804

Sigma-Aldrich

Fe3O4/reduced graphene oxide nanocomposite

10 mg/mL, dispersion in acetone

Synonym(s):

Fe3O4 decorated reduced graphene oxide, Fe3O4/RGO nanocomposite

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.23

description

Loading Density: 30%-70% (Fe3O4 nanocrystal) by TEM
Resistance: <103 Ω/sq (reduced graphene oxide)

form

dispersion

composition

acetone, ~80%
Fe3O4 nanocrystal, 3-8%
reduced graphene oxide, 10-17%

concentration

10 mg/mL

thickness

0.5-1 nm , reduced graphene oxide

particle size

5-25 nm (Fe3O4 nanocrystal)

storage temp.

2-8°C

Application

  • Electromagnetic wave absorption materials.
  • Electro-magnetic device.

Legal Information

Product of Ocean Nanotech, LLC.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

59.0 °F

Flash Point(C)

15 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly efficient dye adsorption and removal: a functional hybrid of reduced graphene oxide-Fe3O4 nanoparticles as an easily regenerative adsorbent.
Zhigang G, et al.
Journal of Materials Chemistry, 22(8), 3527-3535 (2012)
Achieving high specific charge capacitances in Fe3O4/reduced graphene oxide nanocomposites.
Wenhui S, et al.
Journal of Materials Chemistry, 21(10), 3422-3427 (2011)

Articles

Graphene is a unique two-dimensional (2D) structure of monolayer carbon atoms packed into a dense honeycomb crystal that has attracted great interest due to its diverse and fascinating properties.

Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.

Graphene nanoribbons (GNRs) are quasi-one-dimensional narrow strips of graphene comprised of sp2-hybridized carbon atoms arranged into hexagonal honeycomb lattice configurations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service